The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033041.2 | NP_001028213.1 | 1 - 2 | 134 | 74 |
GenBank mRNA | AB071419.1 | - | 1 - 2 | 61 | 74 |
AK143962.1 | - | 1 - 2 | 136 | 74 | |
BC139281.1 | - | 1 - 2 | 85 | 74 |