Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rab3gap1
Entrez Gene ID
226407
Gene Name
RAB3 GTPase activating protein subunit 1
Gene Aliases
1700003B17Rik, 4732493F09Rik, AL117896, Rab3gap, p130
Chromosome Location
Chr.1: 127868737 - 127943876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178690.4 NP_848805.2 11 - 12 1011 90
GenBank mRNA AK076399.1 - 11 - 12 994 90
AK080432.1 - 11 - 12 993 90
AK134604.1 - 11 - 12 993 90
AK164523.1 - 11 - 12 992 90
AK172881.1 - 11 - 12 998 90
BC046297.1 - 11 - 12 977 90

View Full Product Details