Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhdc2
Entrez Gene ID
69554
Gene Name
kelch domain containing 2
Gene Aliases
2310022K15Rik, D12Ertd522e, HCLP-1, LCP
Chromosome Location
Chr.12: 69296681 - 69310687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027117.3 NP_081393.2 2 - 3 543 72
GenBank mRNA AK009481.1 - 2 - 3 458 72
AK051418.1 - 2 - 3 470 72

View Full Product Details