Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc2
Entrez Gene ID
69554
Gene Name
kelch domain containing 2
Gene Aliases
2310022K15Rik, D12Ertd522e, HCLP-1, LCP
Chromosome Location
Chr.12: 69296681 - 69310687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027117.3 NP_081393.2 7 - 8 1029 76
GenBank mRNA AK009481.1 - 7 - 8 944 76
AK164223.1 - 5 - 6 405 76
AY513272.1 - 7 - 8 980 76
BC005581.1 - 7 - 8 953 76

View Full Product Details