Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhdc2
Entrez Gene ID
69554
Gene Name
kelch domain containing 2
Gene Aliases
2310022K15Rik, D12Ertd522e, HCLP-1, LCP
Chromosome Location
Chr.12: 69296681 - 69310687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027117.3 NP_081393.2 12 - 13 1411 106
GenBank mRNA AY513272.1 - 12 - 13 1362 106
BC005581.1 - 12 - 13 1335 106

View Full Product Details