Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhdc2
Entrez Gene ID
69554
Gene Name
kelch domain containing 2
Gene Aliases
2310022K15Rik, D12Ertd522e, HCLP-1, LCP
Chromosome Location
Chr.12: 69296681 - 69310687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY513272.1 - 10 - 11 1223 101
BC005581.1 - 10 - 11 1196 101

View Full Product Details