The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133239.2 | NP_573502.2 | 3 - 4 | 1008 | 80 |
GenBank mRNA | AF406641.1 | - | 3 - 4 | 973 | 80 |
AF492496.1 | - | 3 - 4 | 1032 | 80 | |
AY450552.1 | - | 3 - 4 | 973 | 80 |