The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028185.2 | NP_082461.1 | 3 - 4 | 705 | 97 |
GenBank mRNA | AF514309.1 | - | 3 - 4 | 682 | 97 |
AK008830.1 | - | 1 - 2 | 91 | 97 | |
AK082292.1 | - | 3 - 4 | 706 | 97 | |
AK143661.1 | - | 3 - 4 | 698 | 97 | |
BC094286.1 | - | 3 - 4 | 687 | 97 |