Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp10b
Entrez Gene ID
319767
Gene Name
ATPase, class V, type 10B
Gene Aliases
5930426O13Rik, 9030605H24Rik
Chromosome Location
Chr.11: 42939557 - 43262285 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176999.3 NP_795973.2 12 - 13 2511 72
GenBank mRNA AK220352.1 - 2 - 3 657 72

View Full Product Details