The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029227.1 | NP_083503.1 | 4 - 5 | 421 | 120 |
GenBank mRNA | AK016101.1 | - | 4 - 5 | 406 | 120 |
BC048520.1 | - | 3 - 4 | 377 | 120 |