Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vmn2r1
Entrez Gene ID
56544
Gene Name
vomeronasal 2, receptor 1
Gene Aliases
Casrl1, EG56544, V2r83
Chromosome Location
Chr.3: 64079774 - 64220847 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019918.2 NP_064302.2 4 - 5 1639 139
GenBank mRNA AB267725.1 - 4 - 5 1639 139
AF053986.1 - 4 - 5 1639 139

View Full Product Details