Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zbtb41
Entrez Gene ID
226470
Gene Name
zinc finger and BTB domain containing 41 homolog
Gene Aliases
8430415N23Rik, 9430031N01, 9830132G07Rik, AI316857
Chromosome Location
Chr.1: 139421719 - 139453007 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172643.5 NP_766231.2 10 - 11 2232 129
GenBank mRNA AK034756.1 - 9 - 10 1329 129
AK036544.1 - 10 - 11 2231 129
AK037812.1 - 3 - 4 254 129
AK079719.1 - 6 - 7 590 129
AK163302.1 - 9 - 10 1505 129
BC046465.1 - 9 - 10 1401 129
BC150859.1 - 9 - 10 1571 129
BC172012.1 - 10 - 11 1497 129

View Full Product Details