Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Elf3
Entrez Gene ID
13710
Gene Name
E74-like factor 3
Gene Aliases
ESE-1, ESX, jen
Chromosome Location
Chr.1: 135253574 - 135258565 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163131.1 NP_001156603.1 3 - 4 575 97
NM_007921.3 NP_031947.1 3 - 4 515 97
GenBank mRNA AB221650.1 - 2 - 3 383 97
AF016294.1 - 3 - 4 481 97
AK143135.1 - 3 - 4 565 97
BC138692.1 - 3 - 4 479 97
BC145380.1 - 3 - 4 539 97

View Full Product Details