Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd45
Entrez Gene ID
73844
Gene Name
ankyrin repeat domain 45
Gene Aliases
4933409K03Rik
Chromosome Location
Chr.1: 161142547 - 161170507 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028664.1 NP_082940.1 1 - 2 120 100
GenBank mRNA BC049713.1 - 1 - 2 92 100

View Full Product Details