Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Rap1a
- Entrez Gene ID
-
109905
- Gene Name
- RAS-related protein-1a
- Gene Aliases
-
AI848598, G-22K, Krev-1, Rap1
- Chromosome Location
-
Chr.3: 105727260 - 105801381 on Build GRCm38
- Gene Symbol
- Gm9392
- Entrez Gene ID
-
668838
- Gene Name
- Rap1A-retro1
- Gene Aliases
-
EG668838
- Chromosome Location
-
Chr.7: 46820168 - 46821500 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU359270.1 |
- |
- |
1095 |
138 |
View Full Product Details