The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026958.3 | NP_081234.2 | 3 - 4 | 299 | 102 |
GenBank mRNA | AK007699.1 | - | 3 - 4 | 286 | 102 |
AK028746.1 | - | - | 277 | 102 | |
BC100567.1 | - | 3 - 4 | 264 | 102 | |
BC116739.1 | - | 3 - 4 | 286 | 102 | |
BC116741.1 | - | 3 - 4 | 286 | 102 | |
BU850988.1 | - | 3 - 4 | 302 | 102 | |
CK372908.1 | - | 3 - 4 | 269 | 102 |