Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Adamts20
Entrez Gene ID
223838
Gene Name
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 20
Gene Aliases
Adamst20, bt
Chromosome Location
Chr.15: 94268314 - 94465427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164785.1 NP_001158257.1 4 - 5 1115 64
NM_001164786.1 NP_001158258.1 4 - 5 1115 64
NM_177431.4 NP_803180.3 4 - 5 1115 64
GenBank mRNA AJ512753.3 - 4 - 5 884 64
AK090358.1 - 4 - 5 1081 64
AY189815.1 - 4 - 5 1023 64
AY189816.1 - 4 - 5 1023 64
BC121792.1 - 4 - 5 954 64

View Full Product Details