The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172594.2 | NP_766182.2 | 10 - 11 | 1349 | 71 |
GenBank mRNA | AK049530.1 | - | 10 - 11 | 1422 | 71 |
BC020331.1 | - | 10 - 11 | 1442 | 71 | |
BC057112.1 | - | 10 - 11 | 1422 | 71 | |
BC082319.1 | - | 10 - 11 | 1412 | 71 |