Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ggnbp2
Entrez Gene ID
217039
Gene Name
gametogenetin binding protein 2
Gene Aliases
AI451896, C75939, D330017P12Rik, DIF-3, Zfp403
Chromosome Location
Chr.11: 84832361 - 84870738 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC145605.1 - 5 - 6 764 82
BC150801.1 - 5 - 6 764 82

View Full Product Details