Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fndc1
Entrez Gene ID
68655
Gene Name
fibronectin type III domain containing 1
Gene Aliases
1110027O12Rik, 1110051A18Rik, mKIAA1866
Chromosome Location
Chr.17: 7738568 - 7827342 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081416.1 NP_001074885.1 15 - 16 4683 79

View Full Product Details