Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prame
Entrez Gene ID
75829
Gene Name
preferentially expressed antigen in melanoma
Gene Aliases
4930534P07Rik
Chromosome Location
Chr.X: 135612957 - 135627705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029459.2 NP_083735.2 5 - 6 524 80
GenBank mRNA AK015971.1 - 5 - 6 525 80

View Full Product Details