Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mlc1
Entrez Gene ID
170790
Gene Name
megalencephalic leukoencephalopathy with subcortical cysts 1 homolog (human)
Gene Aliases
AW048630, BB074274, Kiaa0027-hp, LVM, MLC, VL, WKL1, mKIAA0027
Chromosome Location
Chr.15: 88955884 - 88982687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133241.2 NP_573504.1 8 - 9 775 64
GenBank mRNA AF449425.1 - 8 - 9 1229 64
AK030396.1 - 8 - 9 774 64
AK043757.1 - 8 - 9 775 64
AK122190.1 - 8 - 9 1217 64
BC024719.1 - 8 - 9 775 64

View Full Product Details