Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbx2
Entrez Gene ID
21385
Gene Name
T-box 2
Gene Aliases
-
Chromosome Location
Chr.11: 85832615 - 85841948 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009324.2 NP_033350.2 2 - 3 1148 67
GenBank mRNA AK137225.1 - 2 - 3 354 67
AK149348.1 - 2 - 3 956 67
U15566.1 - 2 - 3 684 67

View Full Product Details