The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163102.1 | NP_001156574.1 | 6 - 7 | 127 | |
GenBank mRNA | AK012738.1 | - | 6 - 7 | 126 | |
BC034637.1 | - | - | 127 | ||
BC092228.1 | - | 6 - 7 | 127 |