The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172113.1 | NP_001165584.1 | 1 - 2 | 176 | 74 |
NM_001172114.1 | NP_001165585.1 | 1 - 2 | 176 | 74 | |
NM_153792.2 | NP_722487.2 | 1 - 2 | 176 | 74 | |
GenBank mRNA | AK088431.1 | - | 1 - 2 | 85 | 74 |
AK131617.1 | - | 1 - 2 | 97 | 74 | |
BC008598.1 | - | 1 - 2 | 79 | 74 |