The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028101.4 | NP_082377.2 | 1 - 2 | 120 | 86 |
GenBank mRNA | AK011308.1 | - | 1 - 2 | 117 | 86 |
AK148079.1 | - | 1 - 2 | 98 | 86 | |
AK155810.1 | - | 1 - 2 | 122 | 86 | |
AK172073.1 | - | 1 - 2 | 117 | 86 | |
BC010800.1 | - | 1 - 2 | 82 | 86 |