Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ecel1
Entrez Gene ID
13599
Gene Name
endothelin converting enzyme-like 1
Gene Aliases
DINE, xce
Chromosome Location
Chr.1: 87142874 - 87157085 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277925.1 NP_001264854.1 12 - 13 2009 70
NM_021306.3 NP_067281.2 12 - 13 2001 70
GenBank mRNA AB026294.1 - 11 - 12 1829 70
AK039467.1 - 12 - 13 2002 70
AK159365.1 - 12 - 13 2011 70
BC057569.1 - 11 - 12 1831 70

View Full Product Details