The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146161.1 | NP_001139633.1 | 6 - 7 | 664 | 78 |
NM_008732.2 | NP_032758.2 | 6 - 7 | 664 | 78 | |
GenBank mRNA | AF029758.1 | - | 6 - 7 | 599 | 78 |
AK049856.1 | - | 6 - 7 | 612 | 78 | |
AK083478.1 | - | 6 - 7 | 610 | 78 | |
AK148276.1 | - | 6 - 7 | 612 | 78 | |
AK213433.1 | - | 2 - 3 | 146 | 78 | |
BC019137.1 | - | 6 - 7 | 638 | 78 | |
L33415.1 | - | 6 - 7 | 78 |