The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172587.3 | NP_766175.3 | 1 - 2 | 401 | 130 |
GenBank mRNA | AK044332.1 | - | 1 - 2 | 703 | 130 |
AK161345.1 | - | 1 - 2 | 402 | 130 | |
BC049794.1 | - | 1 - 2 | 343 | 130 | |
BC057357.1 | - | 1 - 2 | 292 | 130 |