The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272056.1 | NP_001258985.1 | 9 - 10 | 1313 | 85 |
NM_010949.2 | NP_035079.1 | 8 - 9 | 1116 | 85 | |
NR_073563.1 | 10 - 11 | 1430 | 85 | ||
GenBank mRNA | AF170709.1 | - | 8 - 9 | 1101 | 85 |
AK004553.1 | - | 10 - 11 | 1338 | 85 | |
BC033459.1 | - | 10 - 11 | 1354 | 85 | |
U70674.1 | - | 9 - 10 | 1234 | 85 |