Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Aatf
- Entrez Gene ID
-
56321
- Gene Name
- apoptosis antagonizing transcription factor
- Gene Aliases
-
4933415H02Rik, 5830465M17Rik, Che-1, Trb
- Chromosome Location
-
Chr.11: 84422856 - 84513570 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK016820.1 |
- |
1 - 2 |
96 |
78 |
View Full Product Details