Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhbdl3
Entrez Gene ID
246104
Gene Name
rhomboid, veinlet-like 3 (Drosophila)
Gene Aliases
AI847581, Rhbdl4, Rhomr1, Vrho
Chromosome Location
Chr.11: 80300432 - 80355986 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139228.3 NP_631974.1 6 - 7 1028 72
GenBank mRNA AJ313479.1 - 6 - 7 1028 72
AK041678.1 - 4 - 5 667 72
BC054784.1 - 6 - 7 864 72
BC056969.1 - 6 - 7 864 72

View Full Product Details