Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Msi2
Entrez Gene ID
76626
Gene Name
musashi RNA-binding protein 2
Gene Aliases
1700105C15Rik, AI451722, AI563628, AW489193, Msi2h
Chromosome Location
Chr.11: 88339382 - 88718267 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_054043.3 NP_473384.1 13 - 14 1217 67
GenBank mRNA AK007122.1 - 3 - 4 426 67
AK049637.1 - 13 - 14 1163 67
AK049688.1 - 13 - 14 1163 67
AK136156.1 - 9 - 10 1025 67
AK153928.1 - 13 - 14 1090 67
AK169846.1 - 13 - 14 1090 67

View Full Product Details