Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hlf
Entrez Gene ID
217082
Gene Name
hepatic leukemia factor
Gene Aliases
E230015K02Rik
Chromosome Location
Chr.11: 90336535 - 90390917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172563.3 NP_766151.1 3 - 4 1222 81
GenBank mRNA AK054062.1 - 3 - 4 712 81
AK079068.1 - 4 - 5 683 81
AK079308.1 - 3 - 4 597 81
BC058705.1 - 3 - 4 1200 81

View Full Product Details