Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zcchc18
Entrez Gene ID
66995
Gene Name
zinc finger, CCHC domain containing 18
Gene Aliases
1500031H04Rik, Sizn2
Chromosome Location
Chr.X: 136992781 - 136996923 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035509.1 NP_001030586.1 3 - 4 443 68
NM_001035510.1 NP_001030587.1 1 - 2 914 68
NM_025893.2 NP_080169.2 3 - 4 391 68
GenBank mRNA AK005317.1 - 3 - 4 369 68
AK134856.1 - - 772 68
BC017627.1 - 3 - 4 304 68
BY261402.1 - 3 - 4 391 68

View Full Product Details