Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Spryd3
Entrez Gene ID
223918
Gene Name
SPRY domain containing 3
Gene Aliases
BC008150
Chromosome Location
Chr.15: 102116526 - 102136245 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033277.3 NP_001028449.2 3 - 4 299 57
GenBank mRNA AK146689.1 - 3 - 4 302 57
BC048820.1 - 3 - 4 276 57
BU057890.1 - 3 - 4 234 57
BY739425.1 - 3 - 4 299 57

View Full Product Details