The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033277.3 | NP_001028449.2 | 10 - 11 | 1250 | 148 |
GenBank mRNA | BC008150.1 | - | 6 - 7 | 761 | 148 |
BC048820.1 | - | 9 - 10 | 1479 | 148 | |
BG293312.1 | - | 3 - 4 | 335 | 148 |