The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199149.3 | NP_954600.1 | 11 - 12 | 2422 | 82 |
GenBank mRNA | AK050946.1 | - | 11 - 12 | 2423 | 82 |
AK160935.1 | - | 11 - 12 | 2426 | 82 | |
BC024868.1 | - | 2 - 3 | 1084 | 82 | |
BC027804.1 | - | 1 - 2 | 166 | 82 | |
BC070402.1 | - | 11 - 12 | 2270 | 82 |