The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199149.3 | NP_954600.1 | 12 - 13 | 2508 | 85 |
GenBank mRNA | AK160935.1 | - | 12 - 13 | 2512 | 85 |
BC024868.1 | - | 3 - 4 | 1170 | 85 | |
BC027804.1 | - | 2 - 3 | 252 | 85 | |
BC070402.1 | - | 12 - 13 | 2356 | 85 |