The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199149.3 | NP_954600.1 | 8 - 9 | 1375 | 66 |
GenBank mRNA | AK050946.1 | - | 8 - 9 | 1376 | 66 |
AK160935.1 | - | 8 - 9 | 1376 | 66 | |
AK190388.1 | - | 1 - 2 | 129 | 66 | |
BC070402.1 | - | 8 - 9 | 1223 | 66 |