Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tfcp2
Entrez Gene ID
21422
Gene Name
transcription factor CP2
Gene Aliases
CP-2, CP2, D230015P20Rik, LBP-1c, LBP-1d, LBP1, LSF, Tcfcp2, UBP-1
Chromosome Location
Chr.15: 100502748 - 100552014 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289603.1 NP_001276532.1 7 - 8 844 86
NM_033476.3 NP_258437.1 7 - 8 844 86
GenBank mRNA AK141248.1 - 1 - 2 184 86
AK146503.1 - 7 - 8 845 86
AK147898.1 - 6 - 7 1053 86
AK158682.1 - 7 - 8 764 86
BC022131.1 - 7 - 8 817 86
M84809.1 - 7 - 8 783 86

View Full Product Details