The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144877.1 | NP_659126.1 | 1 - 2 | 246 | 71 |
GenBank mRNA | AK048636.1 | - | 1 - 2 | 429 | 71 |
AK050492.1 | - | 1 - 2 | 439 | 71 | |
AK084750.1 | - | 1 - 2 | 424 | 71 | |
AK146751.1 | - | - | 444 | 71 | |
AK162843.1 | - | 1 - 2 | 431 | 71 | |
AK168959.1 | - | 1 - 2 | 453 | 71 | |
AK169319.1 | - | - | 445 | 71 | |
AK173044.1 | - | - | 399 | 71 | |
BC014872.1 | - | 1 - 2 | 246 | 71 |