The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144877.1 | NP_659126.1 | 7 - 8 | 1914 | 59 |
GenBank mRNA | AK084750.1 | - | 7 - 8 | 1609 | 59 |
AK146751.1 | - | - | 2112 | 59 | |
AK168959.1 | - | 7 - 8 | 2121 | 59 | |
AK169319.1 | - | - | 2113 | 59 | |
AK173044.1 | - | - | 2067 | 59 | |
BC014872.1 | - | 7 - 8 | 1914 | 59 |