The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011994.2 | NP_036124.2 | 1 - 2 | 1137 | 83 |
GenBank mRNA | AK051445.1 | - | 1 - 2 | 1111 | 83 |
AK087653.1 | - | 1 - 2 | 756 | 83 | |
AK161027.1 | - | 1 - 2 | 798 | 83 | |
BC019187.1 | - | 1 - 2 | 1081 | 83 | |
Z48670.1 | - | 1 - 2 | 1141 | 83 |