Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cwc25
Entrez Gene ID
67480
Gene Name
CWC25 spliceosome-associated protein homolog (S. cerevisiae)
Gene Aliases
1300013D05Rik, AA473636, Ccdc49, R75228
Chromosome Location
Chr.11: 97745470 - 97766613 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026186.4 NP_080462.2 4 - 5 670 124
GenBank mRNA AK004983.2 - 4 - 5 614 124
AK035158.1 - 4 - 5 599 124
BC021478.1 - 4 - 5 584 124
BC079553.1 - 4 - 5 592 124

View Full Product Details