Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jup
Entrez Gene ID
16480
Gene Name
junction plakoglobin
Gene Aliases
Ctnng, D930025P04Rik, PG
Chromosome Location
Chr.11: 100368856 - 100397790 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010593.2 NP_034723.1 11 - 12 2213 82
GenBank mRNA AK145895.1 - 11 - 12 2198 82
AK170934.1 - 11 - 12 2186 82
BC040757.1 - 11 - 12 2162 82
BC094461.1 - 11 - 12 2166 82
M90365.1 - 9 - 10 1678 82

View Full Product Details