Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptgs2
Entrez Gene ID
19225
Gene Name
prostaglandin-endoperoxide synthase 2
Gene Aliases
COX2, Cox-2, PGHS-2, PHS-2, Pghs2, TIS10
Chromosome Location
Chr.1: 150100124 - 150108017 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011198.3 NP_035328.2 2 - 3 275 73
GenBank mRNA AK049923.1 - 2 - 3 266 73
AK144726.1 - 2 - 3 300 73
AK144956.1 - 2 - 3 298 73
AK149820.1 - 2 - 3 300 73
AK166221.1 - 2 - 3 298 73
AK172161.1 - 2 - 3 300 73
BC052900.1 - 2 - 3 271 73
M64291.1 - 2 - 3 299 73
M88242.1 - 2 - 3 275 73
M94967.1 - 2 - 3 300 73

View Full Product Details