Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akap17b
Entrez Gene ID
338351
Gene Name
A kinase (PRKA) anchor protein 17B
Gene Aliases
AA407619, AKAP-17B, B230333C21Rik, C230056F04Rik, PRKA17B, Sfrs17b, Srsf17b, Talia
Chromosome Location
Chr.X: 36608173 - 36645471 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081956.1 NP_001075425.1 4 - 5 1360 134
GenBank mRNA AK136221.1 - 3 - 4 273 134
DV649862.1 - 2 - 3 310 134

View Full Product Details