Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parvg
Entrez Gene ID
64099
Gene Name
parvin, gamma
Gene Aliases
AI413459
Chromosome Location
Chr.15: 84323923 - 84342978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162500.1 NP_001155972.1 6 - 7 676 65
NM_022321.4 NP_071716.3 6 - 7 631 65
GenBank mRNA AF312712.1 - 6 - 7 589 65
AK028329.1 - 6 - 7 632 65
AK037365.1 - 6 - 7 604 65
AK040914.1 - 6 - 7 562 65
AK079881.1 - 6 - 7 557 65
AK088419.1 - 6 - 7 581 65
BC011200.1 - 6 - 7 585 65

View Full Product Details