Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pnpla5
Entrez Gene ID
75772
Gene Name
patatin-like phospholipase domain containing 5
Gene Aliases
4833426H19Rik, GS2L
Chromosome Location
Chr.15: 84112356 - 84127992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029427.1 NP_083703.1 3 - 4 556 103
GenBank mRNA AK014771.1 - 3 - 4 557 103
BC109360.2 - 3 - 4 556 103
BC109361.2 - 3 - 4 556 103

View Full Product Details